Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium acetate 3 M

Supplier: GROWCELLS MS
Synonyms: Acetic acid sodium salt

MRGE-5265-12 MRGE-5265 MRGE-5266-6
101076-100EA 92.63 USD InStock
101076-100 101076-102 101076-162
Sodium acetate 3 M
Sodium acetate
Ready-to-use sterile solution prepared with 18.2 megOhm water, 0.2 micron sterile filtered, pH adjusted to 5.5 with Glacial Acetic acid.


Formula: H₃CCOONa
MW: 82.03 g/mol
MDL Number: MFCD00012459
CAS Number: 127-09-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty