Would you like to log in again?
0.5M EDTA, Grow Cells
Supplier: GrowCells
MRGF-1203 MRGF-1202
101076-178EA 130.79 USD InStock
101076-178 101076-180
0.5M EDTA, Grow Cells
Buffers Cloning Buffers
0.5M Ethylenediaminetetraacetic acid (EDTA)
Prepared with 18.2megOhm water, 0.2 micron filtered and autoclaved in final packaging, pH 8.0.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty