Would you like to log in again?
50X TAE Buffer, Grow Cells
Supplier: GrowCells
MRGF-4210-12 MRGF-4211 MRGF-4211-6 MRGF-4210-010L MRGF-4210
101076-110PK 621.75 USD InStock
101076-110 101076-104 101076-106 101076-216 101076-108
50X TAE Buffer, Grow Cells
Buffers Electrophoresis Buffers Running Buffers
Sterile liquid electrophoresis buffer concentrate prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging.
TAE buffer is used in agarose gel electrophoresis and Polyacrylamide gels both as a buffer and a component of a gel. Can be used for both genomic and large supercoiled DNA. TAE buffer has a relatively low buffering capacity therefore buffer should be replaced periodicly during long electrophoresis runs.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty