Would you like to log in again?
10X TBE, Grow Cells
Supplier: GrowCells
MRGF-4215-12 MRGF-4215
101076-220CS 793.05 USD InStock
101076-220 101076-218
10X TBE, Grow Cells
Buffers Electrophoresis Buffers Running Buffers
Sterile liquid electrophoresis buffer concentrate prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging.
TBE is the most commonly used buffer for DNA and RNA electrophoresis. Since borate is a strong inhibitor of enzymes, TBE is recommended for DNA/RNA sizing, not for recovery of DNA for enzymatic applications. Routinely used for DNA automated sequencing gels. Used both as a running buffer and gel preparation buffer. Recommended for use with fragments smaller than 1500bp
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty