Would you like to log in again?
1M Tris, Grow Cells
Supplier: GrowCells
MRGF-4220 MRGF-4229 MRGF-4229-6 MRGF-4220 CS12
101076-222EA 67.12 USD InStock
101076-222 101076-224 101076-226 101076-228
1M Tris, Grow Cells
Buffers Cloning Buffers
Ready to use, sterile 1.0M Tris buffer solution prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging.
Tris (tris[hydroxymethyl]aminoethane) is widely used as a component of buffer solutions. Suitable for use as an electrophoresis buffer. Tris can be used to increase cell membrane permeability.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty