Would you like to log in again?
1X PBS, Grow Cells
Supplier: GrowCells
MRGF-6230-010L MRGF-6230-12 MRGF-6230 MRGF-6231-6 MRGF-6231
101076-112CS 338.67 USD InStock
101076-112 101076-114 101076-116 101076-238 101076-118
1X PBS, Grow Cells
Buffers Cell Culture Buffers
Ready to use, sterile 1X Phosphate-Buffered Saline solution prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging.
Balanced salt solution used in many biomolecular applications. Ultra pure quality makes buffer ideal for use in a variety of cell culture procedures, such as washing and transporting cells or tissue samples, diluting cell for counting and general preparation of reagents. PBS contains no Ca or Mg ions to provide efficient washing of chelators. Sold in 1X concentration to suit a variety of working dilution for any biological need.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty