Would you like to log in again?
PCR Grade Nuclease Free Water, Grow Cells
Supplier: Grow Cells
PCPW-0125
101385-714PK 34.11 USD InStock
101385-714
PCR Grade Nuclease Free Water, Grow Cells
Sterile PCR grade nuclease free water
Prepared with 18.2 megOhm, 0.1 micron sterile filtered water and autoclaved.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty