Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SP Bel-Art Fritware® Porous Rods, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F13635-0012 F13635-0014
70690-268CS 702.59 USD InStock
70690-268 70690-266
SP Bel-Art Fritware® Porous Rods, Bel-Art Products, a part of SP
Support Rods and Frames
Porous 30.5 cm (12") polyethylene rods can withstand temperatures up to 93°C (200°F).
  • Polyethylene construction
  • Porous design
  • Coarse porosity around 110 microns
Coarse grade porosity with an average pore size of 110 microns.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty