Would you like to log in again?
SP Bel-Art 28-Piece Fitting Kit, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F19626-0000
89009-204PK 119.11 USD InStock
89009-204
SP Bel-Art 28-Piece Fitting Kit, Bel-Art Products, a part of SP
Fittings and Connectors
Compatible with tubing from 3.2 to 12.7 mm (⅛ to ½"), these fittings are for the most common lab connections.
  • Compatible with a wide range of tubing sizes
  • Polypropylene construction
  • Effective organization device
A plastic organizer box has room for adding other fittings and parts. Fittings are molded of autoclavable FDA-grade polypropylene, except the quick disconnect fittings which are polyethylene and not FDA-grade or autoclavable.
Ordering information: The kit contains 6 Straight Tubing Connectors, 7 Stepped Tubing Connectors, 5 Wye (Y) Tubing Connectors, 3 "T" shaped Tubing Connectors, 5 Quick Disconnects and 2 flow control Plastic Tubing Clamps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty