Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-3-hydroxy-1-naphthalenesulfonic acid for analysis of phosphate, Supelco®

Supplier: MilliporeSigma
Synonyms: 1-Amino-2-hydroxy-4-naphthalenesulphonic acid1-Amino-2-hydroxy-4-naphthalenesulfonic acid1-Amino-2-naphthol-4-sulfonic acid4-Amino-3-hydroxy-α-naphthalenesulphonic acid4-Amino-3-hydroxynaphthalene-1-sulphonic acid1-Amino-2-naphthol-4-sulphonic acid4-Amino-3-hydroxy-α-naphthalenesulfonic acid4-Amino-3-hydroxynaphthalene-1-sulfonic acid

1.00099.0025 1.00099.0025
EM1.00099.0025CS 11058 USD InStock
EM1.00099.0025 CA1.00099.0025
4-Amino-3-hydroxy-1-naphthalenesulfonic acid for analysis of phosphate, Supelco®
4-Amino-3-hydroxy-1-naphthalenesulphonic acid


Formula: C₁₀H₉NO₄S
MW: 239.25 g/mol
Boiling Pt: 466 °C (1013 hPa)
Melting Pt: ∼285 °C
Storage Temperature: Ambient
MDL Number: MFCD00004019
CAS Number: 116-63-2
EINECS: 204-147-3
Merck Index: 13,00453
Order Now
Specification Test Results
Solubility (20 °C) practically insoluble
Molar Mass 239.24 g/mol
Bulk Density 280 kg/m3

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty