Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl glyoxime, GR for analysis, reagent for nickel, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: Diacetyl dioxime2,3-Butanedione dioximeDiacetyl glyoxime

1.03062.0100 1.03062.0100
EM1.03062.0100CS 7552 USD InStock
EM1.03062.0100 CA1.03062.0100
Dimethyl glyoxime, GR for analysis, reagent for nickel, Supelco®
Dimethyl glyoxime


Formula: CH₃C(=NOH)C(=NOH)CH₃
MW: 116.12 g/mol
Boiling Pt: 53 °C (8 mmHg)
Melting Pt: 240…241 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002117
CAS Number: 95-45-4
EINECS: 202-420-1
UN: 1325
ADR: 4.1,III
Merck Index: 13,03272
Order Now
Specification Test Results
Solubility 0.6 g/l (20 °C)
Melting Point 239 - 243 °C (decomposition)
Molar Mass 116.11 g/mol
Bulk Density 500 kg/m3
Hazard Class 4.1
Hazard Class Description Flammable solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty