Would you like to log in again?
Ion exchanger(III) (OH⁻ form) Reag. Ph. Eur., GR (Strongly Basic Anion Exchanger), analytical reagent
Supplier: MilliporeSigma
1.04767.0500 1.04767.5000 1.04767.0500 1.04767.5000
52428-638EA 1649.2 USD InStock
CA1.04767.5000 CA1.04767.0500 52428-638 52428-636
Ion exchanger(III) (OH⁻ form) Reag. Ph. Eur., GR (Strongly Basic Anion Exchanger), analytical reagent
Ion exchanger(III) (OH⁻ form)
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Bulk Density 650 - 700 kg/m3
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty