Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dowex® 50WX8 (H) (strongly acidic cation exchanger), Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: Dowex 50 wx8

1.05221.0250 1.05221.0250
52428-652EA 354.8 USD InStock
52428-652 CA1.05221.0250
Dowex® 50WX8 (H) (strongly acidic cation exchanger), Supelco®
Dowex® 50WX8 (H)


MDL Number: MFCD00132726
CAS Number: 11119-67-8
Order Now
Specification Test Results
Description light yellow to brown spherules
Particle size (0,09-0,25 mm) ≥ 70.0 %
Total capacity ≥ 1.7 mol/l

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty