Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dowex® 1X8 (Cl) (strongly basic anion exchanger), Supelco®

Supplier: MilliporeSigma

1.05242.0250 1.05242.0250
52428-658EA 424.43 USD InStock
CA1.05242.0250 52428-658
Dowex® 1X8 (Cl) (strongly basic anion exchanger), Supelco®
Dowex® 1X8 (Cl)


MDL Number: MFCD00132718
CAS Number: 12627-85-9
Order Now
Specification Test Results
Density 1.040 g/cm3 (20 °C)
pH (H2O) not applicable

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty