Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Quinolinol, GR for analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: 8-Hydroxyquinoline

1.07098.0250 1.07098.0250
EM1.07098.0250CS 2567 USD InStock
CA1.07098.0250 EM1.07098.0250
8-Quinolinol, GR for analysis, Supelco®
8-Quinolinol


Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 71.2 °C
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006807
CAS Number: 148-24-3
EINECS: 205-711-1
UN: 2811
ADR: 6.1,II
Merck Index: 13,04869
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/52/770552.jpg
Order Now
Specification Test Results
Solubility 0.56 g/l (20 °C)
Melting Point 72.5 - 74.0 °C
Molar Mass 145.16 g/mol
Boiling Point 267 °C (1013 hPa)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty