Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diatrizoic acid, white

Supplier: CHEM IMPEX INTL INC MS
Synonyms: 3,5-Bis(acetamido)-2,4,6-triiodobenzoic Acid3,5-Di(acetamido)-2,4,6-triiodobenzoic Acid3,5-Diacetamido-2,4,6-triiodobenzoic acid

24150-25G
102601-030EA 68.65 USD InStock
102601-030
Diatrizoic acid, white
Diatrizoic acid
Purified, HPLC.


Formula: C₁₁H₉I₃N₂O₄
MW: 613.92 g/mol
MDL Number: MFCD00069960
CAS Number: 117-96-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty