Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 5 mol/L (5 N) in aqueous solution for molecular biology

Supplier: Promega Corporation
Synonyms: SaltHaliteSea saltRock saltSaline

V4221
PAV4221EA 58.37 USD InStock
PAV4221
Sodium chloride 5 mol/L (5 N) in aqueous solution for molecular biology
Sodium chloride
Used in many molecular biology and forensic applications.

  • Quality tested and certified free of DNase, RNase and protease activity

5M Sodium Chloride is commonly used in many molecular biology and forensic applications.


Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty