Would you like to log in again?
Jacketed Pilot Plant Reactor Pressure Relief Manifold, Ace Glass Incorporated
Supplier: Ace Glass
10015-10
89097-262EA 4371.98 USD InStock
89097-262
Jacketed Pilot Plant Reactor Pressure Relief Manifold, Ace Glass Incorporated
Manifolds
A pressure relief manifold for a jacketed, reaction vessel is designed to protect the vessel from breakage while isolated or disconnected from the circulator
Installed on the outlet side of the vessel,the manifold will reduce the liklihood of breakage caused by an excessive pressure differential exerted by the thermal transfer fluid. Complete system includes aluminum body, outlet fittings, hose, relief valve set at 10 psi, and pressure gauge. Fits 10 l and above.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty