Would you like to log in again?
PTFE Union, Ace Glass Incorporated
Supplier: Ace Glass
12780-04
89077-534EA 76.29 USD InStock
89077-534
PTFE Union, Ace Glass Incorporated
Fittings and Connectors
PTFE union for use with packed columns and solvent delivery systems.
  • Male nipple design
  • 1/4"-28 UNF threads
  • Made of PTFE
PTFE male nipple, 1/4"-28 UNF.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty