Would you like to log in again?
Omnifit Connector Bodies, High Pressure, Ace Glass Incorporated
Supplier: Ace Glass
12741-04 12741-20 12741-40 12741-17 12741-25 12741-14 12741-03 12741-26
89077-490EA 108.55 USD InStock
89077-490 89077-492 89077-480 89077-498 89077-486 89077-488 89077-482 89077-484
Omnifit Connector Bodies, High Pressure, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Ominfit connector bodies are made exclusively of virgin PTFE for maximum chemical inertness.
  • No dead spaces within internal passages
  • Tefzel threaded couplings
  • Rated at 1000psi
  • Autoclavable
The internal passages are 0.8 mm I.D. throughout and there are no dead spaces within. The connectors have 1/4"-28 threaded couplings of Tefzel for instant connection to tube end fittings, plugs or adapters. The connectors are rated at 1000psi and may be autoclaved.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty