Would you like to log in again?
SP Bel-Art Utility Type Funnels, Bel-Art Products, a part of SP
Supplier: Bel-Art
H14781-0000 H14782-0000
70680-065PK 34.54 USD InStock
70680-065 30617-622
SP Bel-Art Utility Type Funnels, Bel-Art Products, a part of SP
Funnels Filtration Funnels Filter Funnels
Externally ribbed polypropylene funnels are steam autoclavable at 121ºC (250ºF) and useful for a variety of lab applications.
  • Polypropylene construction
  • Externally ribbed
  • Autoclavable
Funnels are steam autoclavable at 121ºC (250ºF).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty