Would you like to log in again?
SP Bel-Art Offset Stem Funnel, Bel-Art Products, a part of SP
Supplier: Bel-Art
H14820-0000
70680-042EA 115.39 USD InStock
70680-042
SP Bel-Art Offset Stem Funnel, Bel-Art Products, a part of SP
Funnels Filtration Funnels Filter Funnels
Providing easy access for hard to reach jobs, this funnel features an anti-slop inward flange and large, strong handle.
  • HDPE construction
  • Large, strong handle for easy grip
  • Funnel is chemically resistant
  • Screen is easy to remove and snap back into place
Made of high-density polyethylene plastic, it has a 51 mm (2") diameter, and fine-mesh, bronze filter screen. The screen can be removed with a knife point and easily snapped back into place. The funnel resists most acids, alkalis and some organic solvents. The stem is offset 204 mm (8") center-to-center at a 60 degree angle and tapers to 22 mm (⁷/₈") O.D. Oval top opening is 12.7 x 15.2 cm (5 x 6") with a 10.2 cm (4") deep cone.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty