Would you like to log in again?
Button Type Glass Stirring Shaft, 5 and 6 mm, Ace Glass Incorporated
Supplier: Ace Glass
9534-06 9534-04 9534-40
89075-078EA 78.1 USD InStock
89075-078 89075-080 89075-082
Button Type Glass Stirring Shaft, 5 and 6 mm, Ace Glass Incorporated
Stirrers Stirring Shafts
There is a button on a 5 and 6 mm outer Ø stir rod for the ground glass stirring shaft. This stirring shaft has a total length of 318 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty