Would you like to log in again?
SP Bel-Art CUBE 2UBE™ Rack and Tubes, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F37856-0000 F37853-0001 F37851-1000
20901-949CS 101.5 USD InStock
20901-949 20901-927 47748-184
SP Bel-Art CUBE 2UBE™ Rack and Tubes, Bel-Art Products, a part of SP
Tubes Centrifuge Tubes
CUBE 2UBE™ Tubes offer the advantages and convenience of pipetting 2.2 ml per tube in an 8×12 grid, 96-well format.
  • Polypropylene construction
  • Convenient design
  • Easily fit most automated liquid handling systems
The patented CUBE 2UBE™ Tubes fit most automated liquid handling systems and can be centrifuged up to 14000 rpm.
Made of polypropylene, they can be frozen to –20 °C (–4 °F) and are compatible with Beckman BioMek 1000, Tecan, Hamilton, and other robotic workstations. Each tube has a capacity of 2.2 or 211 ml per rack of 96 tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty