Would you like to log in again?
Heavy Duty Ultrasonic Clamp, Ace Glass Incorporated
Supplier: Ace Glass
9825-21
89075-520EA 739.14 USD InStock
89075-520
Heavy Duty Ultrasonic Clamp, Ace Glass Incorporated
Clamps General Purpose Clamps
For supporting 2-1/2" diameter converter securely in place
Fabricated from 3/4" thick aluminum, anodized black, this clamp fits 1/2" or 5/8" diameter rod and is secured by an Allen head screw to (750W) converter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty