Would you like to log in again?
MMP1
Supplier: Enzo Life Sciences
BML-AK301-0001
10033-750EA 786.73 USD InStock
10033-750
MMP1
Proteins and Peptides
Designed to screen MMP-1 inhibitors using a quenched fluorogenic peptide.
Ordering information: Includes 1 vial MMP-1 enzyme, 1 vial substrate (OmniMMP™ RED), 1 vial 6'-TAMRA calibration standard, 1 vial control inhibitor (NNGH), 1 bottle (20 ml) assay buffer, 1 black 96-well microplate, Instructions.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty