Would you like to log in again?
Ubiquitin-Rhodamine, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-SE761-0025
10033-650EA 253.42 USD InStock
10033-650
Ubiquitin-Rhodamine, Enzo Life Sciences
Proteins and Peptides
Ubiquitin-rhodamine is a high purity industry standard substrate for studying deubiquitinylating activity where detection sensitivity or continuous monitoring of activity.
  • Molecular weight: ~8,9 kDa
It is prepared by the C-terminal derivatisation of ubiquitin with modified rhodamine-110 and has been shown to be a useful and sensitive fluorogenic substrate for wide range of deubiquitinylating enzymes (DUB's), including ubiquitin C-terminal hydrolases (UCH's) and ubiquitin specific proteases (USP's).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty