Would you like to log in again?
Forma™ Audible Temperature Monitors, Thermo Scientific
Supplier: Thermo Scientific
290003
89200-310EA 1534 USD InStock
89200-310
Forma™ Audible Temperature Monitors, Thermo Scientific
Data Loggers
The compact monitor attaches to wall or equipment for convenient single-unit monitoring
For use monitoring temperature of freezers, lab refrigerators, and incubators. Alerts to over- and under-temperature conditions or power loss, with alerm-reset feature. Battery back-up keeps monitor operational, even during a power failure. Ringback system provides additional protection. Self-contained probe. Remote alarm contacts.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty