Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tamoxifen citrate, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: (Z)-1-(4-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate

579000-100MG 579000-100MG
80057-564EA 88.93 USD InStock
80057-564 CA80057-564
Tamoxifen citrate, Sigma-Aldrich®
Tamoxifen citrate
Tamoxifen Citrate is a potent, synthetic, anti-estrogen

Tamoxifen, 4-Hydroxy-, (Z)- is a cell-permeable, active metabolite of Tamoxifen that acts as a potent inhibitor of PKC. It is more potent than the parent compound and inhibits PKC by modifying its catalytic domain.

It is a reversible inhibitor of PKC and induces apoptosis in human malignant glioma cell lines.


Formula: C₂₆H₂₉NO
MW: 563.65 g/mol
MDL Number: MFCD00058321
CAS Number: 54965-24-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty