Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfanilic acid for elementary analysis, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: Aniline-4-sulfonic acid4-Aminobenzenesulfonic acid4-Aminobenzenesulphonic acidAniline-4-sulphonic acid

1.00684.0005 1.00684.0005
EM1.00684.0005CS 3775 USD InStock
CA1.00684.0005 EM1.00684.0005
Sulfanilic acid for elementary analysis, Supelco®
Sulphanilic acid


Formula: 4-(H₂N)C₆H₄SO₃H
MW: 173.19 g/mol
MDL Number: MFCD00007886
CAS Number: 121-57-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/76/05/2297605.jpg
Order Now
Specification Test Results
Solubility 10 g/l (20 °C)
Melting Point 288 °C decomposes
Molar Mass 173.18 g/mol
Bulk Density 620 kg/m3
Density 1.485 g/cm3 (25 °C)
pH 2.5 (10 g/l, H2O, 20 °C)
Ignition Point 400 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty