Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc ≥99.9%, sticks, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

1.08782.0500 1.08782.0500
EM1.08782.0500CS 21058 USD InStock
CA1.08782.0500 EM1.08782.0500
Zinc ≥99.9%, sticks, EMSURE® for analysis, Supelco®
Zinc
Sticks, triangular cross section about 8 mm


Formula: Zn
MW: 65.39 g/mol
MDL Number: MFCD00011291
CAS Number: 7440-66-6
Order Now
Specification Test Results
Assay (complexometric) ≥ 99.9 %
As (Arsenic) ≤ 0.00001 %
Cd (Cadmium) ≤ 0.005 %
Cu (Copper) ≤ 0.002 %
Fe (Iron) ≤ 0.005 %
Pb (Lead) ≤ 0.005 %
Sn (Tin) ≤ 0.001 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty