Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese ≥99%, powder, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

M-Clarity™ quality level = MQ200

1.12237.0250 1.12237.0250
EM1.12237.0250CS 8958 USD InStock
CA1.12237.0250 EM1.12237.0250
Manganese ≥99%, powder, Sigma-Aldrich®
Manganese


Formula: Mn
MDL Number: MFCD00011111
CAS Number: 7439-96-5
UN: 3089
ADR: 4.1,
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 1245 °C
Molar Mass 54.94 g/mol
Boiling Point 2097 °C
Density 7.2 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty