Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium hydrogen citrate sesquihydrate ≥99%, Sigma-Aldrich®

Supplier: MilliporeSigma

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: Sodium hydrogencitrate sesquihydrateDisodium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate sesquihydrateSodium citrate dibasic sesquihydrateCitric acid disodium salt sesquihydrate

M-Clarity™ quality level = MQ200

1.12264.0500 1.12264.0500
EM1.12264.0500CS 1297 USD InStock
EM1.12264.0500 CA1.12264.0500
Disodium hydrogen citrate sesquihydrate ≥99%, Sigma-Aldrich®
di-Sodium hydrogen citrate sesquihydrate


Formula: C₆H₆Na₂O₇·1.5H₂O
MW: 263.11 g/mol
MDL Number: MFCD00150445
CAS Number: 6132-05-4
Order Now
Specification Test Results
Molar Mass 263.11 g/mol

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty