Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diyttrium trioxide, Supelco®

Supplier: MilliporeSigma
Synonyms: Yttrium (III) oxideYttrium oxide

M-Clarity™ quality level = MQ300

1.12412.0025 1.12412.0025
EM1.12412.0025CS 19599 USD InStock
EM1.12412.0025 CA1.12412.0025
Diyttrium trioxide, Supelco®
Diyttrium trioxide


Formula: Y₂O₃
MW: 225.81 g/mol
MDL Number: MFCD00011473
CAS Number: 1314-36-9
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 2410 °C
Molar Mass 225.81 g/mol
Density 5.0 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty