Would you like to log in again?
Iron Test Method: Colorimetric with Color Card and Comparator Block with Long Test Tubes 0.01 - 0.02 - 0.03 - 0.04 - 0.06 - 0.08 - 0.10 - 0.15 -0.20 mg/L Fe MColortest™, MilliporeSigma
Supplier: MilliporeSigma
MColortest™
1.14403.0001 1.14403.0001
EM1.14403.0001CS 2599 USD InStock
CA1.14403.0001 EM1.14403.0001
Iron Test Method: Colorimetric with Color Card and Comparator Block with Long Test Tubes 0.01 - 0.02 - 0.03 - 0.04 - 0.06 - 0.08 - 0.10 - 0.15 -0.20 mg/L Fe MColortest™, MilliporeSigma
Environmental Test Kits
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty