Would you like to log in again?
Chlorine Test (Free Chlorine) Method: Colorimetric, DPD, with Color-Disk Comparator 0.25 - 0.50 - 0.75 - 1 - 2 - 4 - 8 - 10 - 15 mg/L CL₂ MColortest™
Supplier: MilliporeSigma
MColortest™
1.14976.0001 1.14976.0001
EM1.14976.0001CS 7759 USD InStock
EM1.14976.0001 CA1.14976.0001
Chlorine Test (Free Chlorine) Method: Colorimetric, DPD, with Color-Disk Comparator 0.25 - 0.50 - 0.75 - 1 - 2 - 4 - 8 - 10 - 15 mg/L CL₂ MColortest™
Environmental Test Kits
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty