Would you like to log in again?
COD Cell Test for photometers of other manufactures, non Millipore Sigma, Spectroquant®, Millipore Sigma
Supplier: MilliporeSigma
Spectroquant®
1.18752.0007 1.18751.0007
EM1.18752.0007EA 208 USD InStock
EM1.18752.0007 EM1.18751.0007
COD Cell Test for photometers of other manufactures, non Millipore Sigma, Spectroquant®, Millipore Sigma
Environmental Test Kits
Compatible with many photometers and require no programming to measure the tests.
  • 16 mm cells
  • Conforms to ISO 15705
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty