Would you like to log in again?
Chlorine Dioxide Test Method: Colorimetric, DPD, with Color Card and Comparator Block with Long Test Tubes 0.020 - 0.050 - 0.075 - 0.10 - 0.15 - 0.20 - 0.30 - 0.40 - 0.55 mg/L ClO₂ MColortest™
Supplier: MilliporeSigma
MColortest™
1.18754.0001
EM1.18754.0001CS 2976 USD InStock
EM1.18754.0001
Chlorine Dioxide Test Method: Colorimetric, DPD, with Color Card and Comparator Block with Long Test Tubes 0.020 - 0.050 - 0.075 - 0.10 - 0.15 - 0.20 - 0.30 - 0.40 - 0.55 mg/L ClO₂ MColortest™
Environmental Test Kits
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty