Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1H-Pyrazole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 1H-PyrazolePyrazole

M-Clarity™ quality level = MQ200

8.07572.0025 8.07572.0025
EM8.07572.0025CS 6634 USD InStock
EM8.07572.0025 CA8.07572.0025
1H-Pyrazole for synthesis, Sigma-Aldrich®
1H-Pyrazole


Formula: C₃H₄N₂
MW: 68.08 g/mol
MDL Number: MFCD00005234
CAS Number: 288-13-1
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 66 - 68 °C
Molar Mass 68.07 g/mol
Boiling Point 186 - 188 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty