Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methyloctanoic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 4-methylcaprylic acid(R,S)-(+,-)-4-Methyloctanoic acid

M-Clarity™ quality level = MQ200

8.16104.0025 8.16104.0025
EM8.16104.0025CS 37100 USD InStock
CA8.16104.0025 EM8.16104.0025
4-Methyloctanoic acid for synthesis, Sigma-Aldrich®
4-Methylcaprylic acid


Formula: C₉H₁₈O₂
MW: 158.24 g/mol
MDL Number: MFCD00051938
CAS Number: 54947-74-9
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Molar Mass 158.24 g/mol
Boiling Point 292 - 293 °C
Flash Point 99 °C
Density 0.904 - 0.914 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty