Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium osmate(VI) dihydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Potassium dioxidodioxoosmium dihydratePotassium osmium(VI) oxide dihydrateDipotassium osmate dihydrate

M-Clarity™ quality level = MQ200

8.18055.0001 8.18055.0001
EM8.18055.0001CS 263901 USD InStock
CA8.18055.0001 EM8.18055.0001
Potassium osmate(VI) dihydrate for synthesis, Sigma-Aldrich®
Potassium osmate(VI) dihydrate


Formula: K₂O₄Os·2H₂O
MW: 368.45 g/mol
MDL Number: MFCD00149919
CAS Number: 10022-66-9
UN: 3085
ADR: 5.1 (8),II
Order Now
Specification Test Results
Solubility (20 °C) soluble
Molar Mass 368.43 g/mol

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty