Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylphosphonic dichloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Dichlorophenylphosphine oxidePhenyldichlorophosphine oxide

M-Clarity™ quality level = MQ200

8.18248.0100 8.18248.0100
EM8.18248.0100CS 1644 USD InStock
EM8.18248.0100 CA8.18248.0100
Phenylphosphonic dichloride for synthesis, Sigma-Aldrich®
Phenylphosphonic dichloride


Formula: C₆H₅Cl₂OP
MW: 194.98 g/mol
MDL Number: MFCD00002070
CAS Number: 824-72-6
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Solubility (20 °C) (decomposition)
Melting Point 1 °C
Molar Mass 194.98 g/mol
Boiling Point 258 °C
Flash Point 204 °C
Refractive Index 1.5581 (25 °C, 589 nm)
Density 1.40 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty