Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium iodide, anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

8.18287.0050 8.18287.0050
EM8.18287.0050CS 15950 USD InStock
EM8.18287.0050 CA8.18287.0050
Lithium iodide, anhydrous for synthesis, Sigma-Aldrich®
Lithium iodide


Formula: ILi
MW: 133.85 g/mol
MDL Number: MFCD00011092
CAS Number: 10377-51-2
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 450 °C
Molar Mass 133.84 g/mol
Bulk Density 1000 kg/m3
Boiling Point 1180 °C
Density 4 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty