Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexamethyldisilane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

8.18889.0050 8.18889.0010 8.18889.0010 8.18889.0050
EM8.18889.0010CS 70083 USD InStock
CA8.18889.0010 EM8.18889.0010 CA8.18889.0050 EM8.18889.0050
Hexamethyldisilane for synthesis, Sigma-Aldrich®
Hexamethyldisilane


Formula: C₆H₁₈Si₂
MDL Number: MFCD00008258
CAS Number: 1450-14-2
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Solubility (20 °C) (decomposition)
Melting Point 13 - 15 °C
Molar Mass 146.37 g/mol
Boiling Point 110 - 112 °C
Flash Point -6 °C
Refractive Index 1.4207 (20 °C, 589 nm)
Density 0.73 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty