Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-N,N-diethylacetamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 2-Chloro-n,n-diethylacetamideN,N,-Diethylchloroacetamide

M-Clarity™ quality level = MQ200

8.20399.0025 8.20399.0025
EM8.20399.0025CS 10896 USD InStock
CA8.20399.0025 EM8.20399.0025
2-Chloro-N,N-diethylacetamide for synthesis, Sigma-Aldrich®
2-Chloro-N,N-diethylacetamide


Formula: C₆H₁₂ClNO
MW: 149.62 g/mol
MDL Number: MFCD00000928
CAS Number: 2315-36-8
UN: 2810
ADR: 6.1,II
Order Now
Specification Test Results
Solubility 85.5 g/l (25 °C)
Molar Mass 149.61 g/mol
Boiling Point 114 - 116 °C (23 hPa)
Flash Point 113 °C
Refractive Index 1.469 (21 °C, 589 nm)
Density 1.09 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty