Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethyl-4-nitrosoaniline for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 4-Nitroso-N,N-dimethylaniline

M-Clarity™ quality level = MQ200

8.20515.0025 8.20515.0025
EM8.20515.0025CS 1453 USD InStock
EM8.20515.0025 CA8.20515.0025
N,N-Dimethyl-4-nitrosoaniline for synthesis, Sigma-Aldrich®
N,N-Dimethyl-4-nitrosoaniline


Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
MDL Number: MFCD00002063
CAS Number: 138-89-6
UN: 1369
ADR: 4.2,II
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Melting Point 84 - 87 °C
Molar Mass 150.18 g/mol
Density 1.15 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty