Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

O-Benzylhydroxylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

8.20617.0010 8.20617.0010
EM8.20617.0010CS 123056 USD InStock
EM8.20617.0010 CA8.20617.0010
O-Benzylhydroxylamine for synthesis, Sigma-Aldrich®
O-Benzylhydroxylamine


Formula: C₇H₉NO
MDL Number: MFCD00221709
CAS Number: 622-33-3
UN: 2735
ADR: 8,II
Order Now
Specification Test Results
Solubility 47 g/l (15 °C)
Molar Mass 123.16 g/mol
Boiling Point 49 °C (1 hPa)
Vapor Pressure 2.09 hPa (58 °C)
Flash Point 91.5 °C
Density 1.064 g/cm3 (23 °C)
pH 8 (47 g/l, H2O)
Ignition Point 260 °C DIN 51794
Viscosity Kinematic 3.34 mm2/s (23 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty