Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

1,3,5-Trioxane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: sym. Trioxanes-TrioxaneTrioxymethyleneFormaldehyde trimer

M-Clarity™ quality level = MQ200

8.21189.0100 8.21189.1000 8.21189.9025 8.21189.1000 8.21189.0100 8.21189.9025
EM8.21189.9025EA 1298 USD InStock
EM8.21189.9025 EM8.21189.0100 EM8.21189.1000 CA8.21189.9025 CA8.21189.1000 CA8.21189.0100
1,3,5-Trioxane for synthesis, Sigma-Aldrich®
1,3,5-Trioxane


Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 114.5 °C (1013 hPa)
Melting Pt: 60…62 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00006563
CAS Number: 110-88-3
EINECS: 203-812-5
UN: 1325
ADR: 4.1,III
Merck Index: 12,09863
Order Now
Specification Test Results
Solubility 221 g/l (25 °C)
Melting Point 60 - 62 °C
Molar Mass 90.08 g/mol
Boiling Point 114 - 115 °C (1013 hPa)
Vapor Pressure 7.5 hPa (20 °C)
Flash Point 45 °C
Explosion Limit 3.6 - 28.7 %(V)
Density 1.17 g/cm3 (65 °C)
pH (H2O) neutral
Ignition Point 410 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty