Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Lithium oxalate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: Lithium oxalateDilithium oxalateLithiumoxalate

M-Clarity™ quality level = MQ200

8.22085.0100 8.22085.0100
EM8.22085.0100CS 2214 USD InStock
CA8.22085.0100 EM8.22085.0100
di-Lithium oxalate for synthesis, Sigma-Aldrich®
di-Lithium oxalate


Formula: C₂Li₂O₄
MW: 101.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040596
CAS Number: 553-91-3
EINECS: 209-054-1
ADR: 6.1,III
Order Now
Specification Test Results
Solubility 66 g/l (20 °C)
Molar Mass 101.9 g/mol

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty