Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Platinum(II) chloride Pt 73% for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Platinum dichloride

M-Clarity™ quality level = MQ200

8.24566.0500 8.24566.0500
EM8.24566.0500CS 685422 USD InStock
CA8.24566.0500 EM8.24566.0500
Platinum(II) chloride Pt 73% for synthesis, Sigma-Aldrich®
Platinum (II) chloride


Formula: Cl₂Pt
MW: 265.99 g/mol
Melting Pt: 581 °C
Density: 6.05 g/cm³ (20 °C)
MDL Number: MFCD00011181
CAS Number: 10025-65-7
EINECS: 233-034-1
UN: 1759
ADR: 8,III
Merck Index: 12,07687
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 581 °C decomposes
Molar Mass 265.99 g/mol
Density 6.1 g/cm3
pH 5 - 6 (H2O) suspension

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty